Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRPC7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001167576.1 | NP_001161048.1 | 3 | 1069 | ||
NM_001167577.1 | NP_001161049.1 | 4 | 1234 | |||
NM_020389.2 | NP_065122.1 | 5 | 1417 | |||
GenBank mRNA | AJ272034.1 | - | 4 | 1195 | ||
AJ421783.1 | - | 4 | 1195 | |||
AJ549088.1 | - | 3 | 1012 | |||
AJ549089.1 | - | 3 | 1030 | |||
AY167926.1 | - | 3 | 1027 | |||
AY167927.1 | - | 2 | 847 | |||
BC128185.1 | - | 4 | 1195 | 40113370 | ||
BC128186.1 | - | 4 | 1211 |