Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CNTFR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001207011.1 | NP_001193940.1 | 6 | 704 | ||
NM_001842.4 | NP_001833.1 | 5 | 717 | |||
NM_147164.2 | NP_671693.1 | 6 | 828 | |||
XM_017014260.1 | XP_016869749.1 | 7 | 1044 | |||
XM_017014261.1 | XP_016869750.1 | 7 | 1017 | |||
XM_017014262.1 | XP_016869751.1 | 7 | 961 | |||
XM_017014263.1 | XP_016869752.1 | 6 | 802 | |||
XM_017014264.1 | XP_016869753.1 | 6 | 1346 | |||
XM_017014265.1 | XP_016869754.1 | 5 | 662 | |||
GenBank mRNA | AK127444.1 | - | 2 | 1809 | ||
AL537375.3 | - | 6 | 703 | |||
BC001492.2 | - | 5 | 674 | 3510004 | ||
BT019824.1 | - | 4 | 534 | |||
HQ628632.1 | - | 4 | 558 | |||
M73238.1 | - | 6 | 797 |