Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD177 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020406.3 | NP_065139.2 | 6 | 753 | ||
XM_017027021.1 | XP_016882510.1 | 6 | 737 | |||
XM_017027022.1 | XP_016882511.1 | 6 | 737 | |||
GenBank mRNA | AB237911.1 | - | 6 | 705 | ||
AF146747.1 | - | 6 | 732 | |||
AJ290452.1 | - | 6 | 724 | |||
AJ305326.1 | - | 6 | 842 | |||
AJ310433.1 | - | 7 | 843 | |||
AK225226.1 | - | 6 | 726 | |||
AK290965.1 | - | 6 | 728 | |||
AY358932.1 | - | 6 | 713 | |||
BC029167.1 | - | 6 | 715 | 5182826 | ||
BT020111.1 | - | 6 | 697 | |||
CD514938.1 | - | 6 | 701 | |||
DA728784.1 | - | 6 | 751 |