Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LYRM4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164841.2 | NP_001158313.1 | 4 | 1176 | ||
NM_020408.5 | NP_065141.3 | 3 | 1051 | |||
NR_104417.1 | 4 | 1398 | ||||
NR_104418.1 | 2 | 930 | ||||
GenBank mRNA | AF258559.1 | - | 1 | 370 | ||
AK309458.1 | - | 3 | 1097 | |||
BC067762.1 | - | 2 | 722 |