Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: MRPL47 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020409.2 | NP_065142.2 | 6 | 608 | ||
NM_177988.1 | NP_817125.1 | 5 | 462 | |||
GenBank mRNA | AY212270.1 | - | 6 | 585 | ||
BC021575.1 | - | 5 | 462 | 3161966 | ||
BC032522.1 | - | 6 | 541 | 5500168 | ||
CA428787.1 | - | 6 | 621 |