Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RGL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001035223.3 | NP_001030300.3 | 8, 9 | 1172 | ||
NM_001161616.2 | NP_001155088.2 | 8, 9 | 1172 | |||
GenBank mRNA | AK057147.1 | - | 9, 10 | 1327 | ||
AK096811.1 | - | 4, 5 | 489 | |||
AK293404.1 | - | 7, 8 | 899 | |||
BC014426.1 | - | 8, 9 | 1158 | |||
BC101756.1 | - | 8, 9 | 1158 | 8069262 | ||
BC111958.1 | - | 8, 9 | 1158 | 8327426 | ||
BC143530.1 | - | 8, 9 | 1158 | 9052039 |