Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other RTN4 SIRNA Assays ›
Gene Symbol:
RTN4
Gene Name:
reticulon 4
Gene Aliases:
ASY, NI220/250, NOGO, NOGO-A, NOGOC, NSP, NSP-CL, Nbla00271, Nbla10545, Nogo-B, Nogo-C, RTN-X, RTN4-A, RTN4-B1, RTN4-B2, RTN4-C
Chromosome Location:
Chr.2: 54972187 - 55137831 on Build GRCh38
UniGene ID:
Hs.637850
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s32766
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: RTN4
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001321859.1
NP_001308788.1
5
2876
NM_001321860.1
NP_001308789.1
5
2946
NM_001321861.1
NP_001308790.1
5
2909
NM_001321862.1
NP_001308791.1
5
2896
NM_001321863.1
NP_001308792.1
5
2846
NM_001321904.1
NP_001308833.1
6
3324
NM_007008.2
NP_008939.1
3
566
NM_020532.4
NP_065393.1
5
3629
NM_153828.2
NP_722550.1
3
1172
NM_207520.1
NP_997403.1
4
1229
NM_207521.1
NP_997404.1
5
2887
NR_135829.1
3
452
NR_135830.1
3
430
XM_005264434.3
XP_005264491.1
6
3023
XM_017004518.1
XP_016860007.1
6
3062
XM_017004519.1
XP_016860008.1
6
3179
GenBank mRNA
AB015639.1
-
3
940
AB020693.1
-
5
3465
AB040462.1
-
5
3575
AB040463.1
-
3
1118
AB073351.1
-
3
1301
AF063601.1
-
3
1889
AF077050.1
-
3
565
AF087901.1
-
3
457
AF125103.1
-
3
566
AF132047.1
-
3
1053
AF132048.1
-
3
472
AF148537.1
-
5
3472
AF148538.1
-
3
1015
AF177332.1
-
3
471
AF320999.1
-
4
2635
AF333336.1
-
5
2891
AJ251383.1
-
5
3331
AJ251384.1
-
3
874
AJ251385.1
-
3
352
AK075039.1
-
3
1106
AK129806.1
-
3
1031
AK130812.1
-
3
933
AY102276.1
-
3
586
AY102277.1
-
3
1118
AY102278.1
-
4
1175
AY102279.1
-
5
3575
AY123245.1
-
5
2888
AY123246.1
-
5
2946
AY123247.1
-
5
2909
AY123248.1
-
5
2895
AY123249.1
-
5
2856
AY123250.1
-
5
2846
BC001035.2
-
3
537
BC007109.1
-
3
463
BC010737.1
-
3
879
BC012619.1
-
3
1029
BC014366.1
-
3
463
BC016165.1
-
4
1178
BC026788.1
-
3
1060
BC068991.1
-
3
1122
BC071848.1
-
4
860
BC150182.1
-
5
3579
40148210
BC152425.1
-
5
3465
8862539
BC152555.1
-
5
3579
40148984
CN429712.1
-
3
401
DQ778739.1
-
3
352
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
apoptotic process
axonal fasciculation
cerebral cortex radial glia guided migration
negative regulation of cell growth
negative regulation of axon extension
regulation of apoptotic process
negative regulation of axonogenesis
nuclear pore complex assembly
cardiac epithelial to mesenchymal transition
endoplasmic reticulum tubular network organization
endoplasmic reticulum tubular network assembly
cell-cell adhesion
regulation of branching morphogenesis of a nerve
protein binding
poly(A) RNA binding
cadherin binding involved in cell-cell adhesion
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7c5f844ff9-7cf5v:80/10.239.163.60:80.
git-commit: 1d07a45fbd3445ee17db916b8ffd5a90ab36fdd0
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.11.0-2024.07.28-1.0