Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KIAA1191 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001079684.2 | NP_001073152.1 | 6 | 762 | ||
NM_001079685.2 | NP_001073153.1 | 6 | 741 | |||
NM_001287335.1 | NP_001274264.1 | 4 | 475 | |||
NM_001287336.1 | NP_001274265.1 | 5 | 654 | |||
NM_020444.4 | NP_065177.2 | 7 | 849 | |||
NR_109796.1 | 3 | 343 | ||||
NR_109797.1 | 3 | 310 | ||||
NR_109798.1 | 4 | 456 | ||||
NR_109799.1 | 4 | 418 | ||||
NR_109800.1 | 3 | 331 | ||||
XM_005265941.1 | XP_005265998.1 | 6 | 662 | |||
XM_005265945.1 | XP_005266002.1 | 4 | 337 | |||
XM_005265946.4 | XP_005266003.1 | 6 | 757 | |||
XM_011534595.1 | XP_011532897.1 | 6 | 624 | |||
XM_011534596.1 | XP_011532898.1 | 5 | 573 | |||
XM_017009651.1 | XP_016865140.1 | 5 | 558 | |||
XM_017009652.1 | XP_016865141.1 | 2 | 281 | |||
XM_017009653.1 | XP_016865142.1 | 4 | 430 | |||
GenBank mRNA | AK315428.1 | - | 7 | 806 | ||
AL390217.1 | - | 6 | 720 | |||
AL832558.1 | - | 1 | 572 | |||
BC006316.2 | - | 3 | 263 | |||
BC010448.1 | - | 7 | 784 | 4155679 | ||
BC012848.1 | - | 4 | 353 | |||
BC017097.1 | - | 7 | 806 | 3529175 | ||
BM547296.1 | - | 3 | 291 | |||
BX648859.1 | - | 7 | 848 | |||
CN341192.1 | - | 4 | 425 | |||
CR457331.1 | - | 5 | 476 | |||
DQ073392.1 | - | 5 | 476 | |||
HY114499.1 | - | 4 | 433 | |||
HY120613.1 | - | 3 | 330 |