Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SEPN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020451.2 | NP_065184.2 | 4 | 569 | ||
NM_206926.1 | NP_996809.1 | 3 | 467 | |||
GenBank mRNA | AA613025.1 | - | 2 | 189 | ||
AF166125.1 | - | 2 | 158 | |||
AJ306399.1 | - | 4 | 569 | |||
AK308457.1 | - | 2 | 188 |