Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SEPN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020451.2 | NP_065184.2 | 10 | 1349 | ||
NM_206926.1 | NP_996809.1 | 9 | 1247 | |||
GenBank mRNA | AF166125.1 | - | 8 | 938 | ||
AJ306399.1 | - | 10 | 1349 | |||
AK308457.1 | - | 8 | 967 | |||
BC015638.2 | - | 1 | 14 | |||
BC021028.2 | - | 5 | 552 | |||
BC042154.2 | - | 1 | 14 |