Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATP10A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024490.3 | NP_077816.1 | 16 | 3294 | ||
XM_005268261.4 | XP_005268318.1 | 17 | 3424 | |||
XM_011521826.2 | XP_011520128.1 | 17 | 3595 | |||
XM_011521828.2 | XP_011520130.1 | 16 | 4023 | |||
XM_011521829.2 | XP_011520131.1 | 13 | 2427 | |||
XM_011521830.2 | XP_011520132.2 | 11 | 2097 | |||
XM_017022436.1 | XP_016877925.1 | 16 | 2930 | |||
XM_017022437.1 | XP_016877926.1 | 16 | 3294 | |||
GenBank mRNA | AB011138.1 | - | 11 | 2182 | ||
AB051358.1 | - | 17 | 3387 | |||
BC052251.1 | - | 16 | 3300 | 6258931 |