Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: COL4A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001303110.1 | NP_001290039.1 | 3, 4 | 372 | ||
NM_001845.5 | NP_001836.3 | 3, 4 | 372 | |||
XM_011521048.2 | XP_011519350.1 | 3, 4 | 8304 | |||
GenBank mRNA | AB209646.1 | - | 3, 4 | 357 | ||
BC142626.1 | - | 3, 4 | 346 | |||
BC151220.1 | - | 3, 4 | 356 | 40148649 | ||
CD613007.1 | - | 19 | ||||
CN256264.1 | - | 3, 4 | 365 | |||
X05561.1 | - | 3, 4 | 364 | |||
Y00706.1 | - | 1, 2 | 107 |