Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other SNX14 SIRNA Assays ›
Gene Symbol:
SNX14
Gene Name:
sorting nexin 14
Gene Aliases:
RGS-PX2, SCAR20
Chromosome Location:
Chr.6: 85505496 - 85594156 on Build GRCh38
UniGene ID:
Hs.485871
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s32929
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: SNX14
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001297614.2
NP_001284543.1
18
2244
NM_001304479.1
NP_001291408.1
19
2016
NM_020468.5
NP_065201.1
16
2112
NM_153816.5
NP_722523.1
19
2271
XM_005248738.3
XP_005248795.1
19
2244
XM_005248740.3
XP_005248797.1
17
2113
XM_011535977.2
XP_011534279.1
18
1687
XM_011535979.1
XP_011534281.1
17
1679
XM_017011090.1
XP_016866579.1
19
1753
XM_017011091.1
XP_016866580.1
18
2217
XM_017011092.1
XP_016866581.1
17
1881
XM_017011093.1
XP_016866582.1
19
1958
XM_017011094.1
XP_016866583.1
17
1853
XM_017011095.1
XP_016866584.1
17
1593
XM_017011096.1
XP_016866585.1
19
1982
XM_017011097.1
XP_016866586.1
19
1966
XM_017011098.1
XP_016866587.1
18
1884
XM_017011099.1
XP_016866588.1
17
1850
XM_017011100.1
XP_016866589.1
16
1574
XM_017011101.1
XP_016866590.1
17
1656
GenBank mRNA
AF121863.1
-
2
117
AK000362.1
-
19
1960
AK026479.1
-
17
2376
AK095380.1
-
19
2440
AK125063.1
-
15
2323
AK226097.1
-
7
582
AK295417.1
-
19
1745
AY044865.1
-
16
1653
BC005110.1
-
16
1841
3959086
BC046520.2
-
18
1699
BC068589.1
-
19
2010
4818366
BC095419.1
-
19
1856
5268442
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: NM_001108174.
Panther Classification:
Molecular Function -
scaffold/adaptor protein
Gene Ontology Categories:
Function(s)
Process(es)
protein transport
autophagosome maturation
phosphatidylinositol binding
phosphatidylinositol-3,5-bisphosphate binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5d97d974fc-xql29:80/10.239.165.153:80.
git-commit: fe8b04579249ff98075bad6061a6e2a4739e01ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.13.0-2024.08.32-1.0