Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RCN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020650.2 | NP_065701.2 | 5, 6 | 1126 | ||
XM_005259089.3 | XP_005259146.1 | 6, 7 | 1221 | |||
XM_011527143.1 | XP_011525445.1 | 6, 7 | 1315 | |||
XM_017027023.1 | XP_016882512.1 | 5, 6 | 1400 | |||
GenBank mRNA | AF183423.1 | - | 4, 5 | 711 | ||
AY195859.1 | - | 5, 6 | 1126 | |||
AY358423.1 | - | 5, 6 | 713 | |||
BC013436.1 | - | 5, 6 | 709 | 3889718 |