Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SENP7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077203.2 | NP_001070671.1 | 9 | 1313 | ||
NM_001282801.1 | NP_001269730.1 | 9 | 1310 | |||
NM_001282802.1 | NP_001269731.1 | 9 | 1458 | |||
NM_001282803.1 | NP_001269732.1 | 7 | 1065 | |||
NM_020654.4 | NP_065705.3 | 10 | 1508 | |||
XM_005247662.4 | XP_005247719.1 | 8 | 1101 | |||
XM_011513038.2 | XP_011511340.1 | 10 | 1497 | |||
XM_011513039.2 | XP_011511341.1 | 9 | 1422 | |||
XM_011513040.2 | XP_011511342.1 | 9 | 1302 | |||
XM_011513041.2 | XP_011511343.1 | 9 | 1299 | |||
XM_011513042.2 | XP_011511344.1 | 8 | 1104 | |||
XM_011513045.2 | XP_011511347.1 | 10 | 1498 | |||
XM_017006924.1 | XP_016862413.1 | 10 | 1385 | |||
XM_017006925.1 | XP_016862414.1 | 10 | 1473 | |||
XM_017006926.1 | XP_016862415.1 | 8 | 1211 | |||
XM_017006927.1 | XP_016862416.1 | 8 | 1208 | |||
XM_017006928.1 | XP_016862417.1 | 10 | 1498 | |||
GenBank mRNA | AB051494.1 | - | 3 | 515 | ||
AF199458.1 | - | 9 | 1456 | |||
AK297356.1 | - | 1419 | ||||
AL136599.1 | - | 9 | 1336 | |||
AL833577.1 | - | 9 | 1279 | |||
BC129988.1 | - | 10 | 1435 | 40128878 | ||
BX537943.1 | - | 7 | 1009 |