Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SENP7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077203.2 | NP_001070671.1 | 6 | 613 | ||
NM_001282801.1 | NP_001269730.1 | 6 | 610 | |||
NM_001282802.1 | NP_001269731.1 | 6 | 758 | |||
NM_001282803.1 | NP_001269732.1 | 4 | 365 | |||
NM_001282804.1 | NP_001269733.1 | 5 | 563 | |||
NM_020654.4 | NP_065705.3 | 7 | 808 | |||
XM_005247662.4 | XP_005247719.1 | 5 | 401 | |||
XM_011513038.2 | XP_011511340.1 | 7 | 797 | |||
XM_011513039.2 | XP_011511341.1 | 6 | 722 | |||
XM_011513040.2 | XP_011511342.1 | 6 | 602 | |||
XM_011513041.2 | XP_011511343.1 | 6 | 599 | |||
XM_011513042.2 | XP_011511344.1 | 5 | 404 | |||
XM_011513045.2 | XP_011511347.1 | 7 | 798 | |||
XM_017006924.1 | XP_016862413.1 | 7 | 685 | |||
XM_017006925.1 | XP_016862414.1 | 7 | 773 | |||
XM_017006926.1 | XP_016862415.1 | 5 | 511 | |||
XM_017006927.1 | XP_016862416.1 | 5 | 508 | |||
XM_017006928.1 | XP_016862417.1 | 7 | 798 | |||
XM_017006929.1 | XP_016862418.1 | 6 | 602 | |||
GenBank mRNA | AF199458.1 | - | 6 | 756 | ||
AK297356.1 | - | 719 | ||||
AL136599.1 | - | 6 | 636 | |||
AL833577.1 | - | 6 | 579 | |||
BC062674.1 | - | 7 | 755 | |||
BC129988.1 | - | 7 | 735 | 40128878 | ||
BX537943.1 | - | 4 | 309 |