Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: AS3MT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020682.3 | NP_065733.2 | 4 | 337 | ||
GenBank mRNA | AF226730.1 | - | 4 | 274 | ||
AK057833.1 | - | 4 | 356 | |||
AK225972.1 | - | 4 | 356 | |||
AK309737.1 | - | 4 | 269 | |||
BC119637.1 | - | 4 | 241 | |||
BC119638.2 | - | 4 | 241 | |||
DA368970.1 | - | 4 | 337 |
Gene Symbol: BORCS7-ASMT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_037644.1 | 8 | 602 |