Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMCC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301036.1 | NP_001287965.1 | 2 | 297 | ||
NM_020698.3 | NP_065749.2 | 2 | 278 | |||
GenBank mRNA | AB032971.1 | - | 2 | 207 | ||
BC040535.1 | - | 2 | 264 | 5264859 |