Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC12A5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134771.1 | NP_001128243.1 | 25 | 3390 | ||
NM_020708.4 | NP_065759.1 | 25 | 3408 | |||
XM_017027981.1 | XP_016883470.1 | 24 | 3375 | |||
GenBank mRNA | AB033002.1 | - | 25 | 3223 | ||
AF208159.1 | - | 25 | 3245 | |||
AK289758.1 | - | 25 | 3367 | |||
AK294059.1 | - | 21 | 2585 | |||
AK295096.1 | - | 9 | 950 | |||
AK295900.1 | - | 19 | 2604 | |||
BC132668.1 | - | 25 | 3333 | 40146690 | ||
BC132670.1 | - | 25 | 3333 | 40146692 |