Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLEKHG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001029884.1 | NP_001025055.1 | 6 | 926 | ||
XM_005267064.3 | XP_005267121.1 | 5 | 3005 | |||
XM_005267065.3 | XP_005267122.1 | 7 | 952 | |||
XM_005267066.2 | XP_005267123.1 | 5 | 1809 | |||
XM_006715521.1 | XP_006715584.1 | 7 | 1179 | |||
XM_006715522.3 | XP_006715585.1 | 5 | 3046 | |||
XM_011535981.2 | XP_011534283.1 | 5 | 3802 | |||
XM_011535982.2 | XP_011534284.1 | 5 | 3802 | |||
XM_011535983.2 | XP_011534285.1 | 6 | 2227 | |||
XM_017011102.1 | XP_016866591.1 | 5 | 1811 | |||
GenBank mRNA | AJ420468.1 | - | 3 | 164 | ||
AK056300.1 | - | 5 | 825 | |||
AK293288.1 | - | 5 | 857 | |||
AK294798.1 | - | 5 | 980 | |||
AK307631.1 | - | 5 | 879 | |||
BC089428.1 | - | 1 | 37 | |||
BC140864.1 | - | 4 | 663 | 9021732 | ||
BC151134.1 | - | 4 | 663 | 9057034 |