Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARID1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017519.2 | NP_059989.2 | 4 | 1907 | ||
NM_020732.3 | NP_065783.3 | 5 | 1946 | |||
XM_005267069.4 | XP_005267126.2 | 4 | 1982 | |||
XM_011535984.2 | XP_011534286.2 | 5 | 2022 | |||
XM_017011103.1 | XP_016866592.1 | 5 | 2022 | |||
XM_017011104.1 | XP_016866593.1 | 5 | 2021 | |||
XM_017011105.1 | XP_016866594.1 | 5 | 2022 | |||
XM_017011106.1 | XP_016866595.1 | 5 | 2021 | |||
XM_017011107.1 | XP_016866596.1 | 5 | 2022 | |||
XM_017011108.1 | XP_016866597.1 | 5 | 2029 | |||
GenBank mRNA | AF253515.1 | - | 4 | 1101 | ||
AF468300.1 | - | 5 | 476 | |||
AF521671.1 | - | 4 | 1694 | |||
AK025945.1 | - | 2 | 173 | |||
AK074180.1 | - | 1 | 15 |