Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COL6A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004369.3 | NP_004360.2 | 41 | 9503 | ||
NM_057166.4 | NP_476507.3 | 38 | 7682 | |||
NM_057167.3 | NP_476508.2 | 40 | 8885 | |||
XM_005246065.1 | XP_005246122.1 | 40 | 8860 | |||
XM_005246066.1 | XP_005246123.1 | 38 | 8006 | |||
XM_006712253.1 | XP_006712316.1 | 41 | 8959 | |||
XM_011510574.1 | XP_011508876.1 | 41 | 9457 | |||
XM_017003303.1 | XP_016858792.1 | 46 | 9510 | |||
XM_017003304.1 | XP_016858793.1 | 43 | 7722 | |||
GenBank mRNA | AB209636.1 | - | 32 | 4794 | ||
BC033174.1 | - | 18 | 2626 | |||
BC144595.1 | - | 38 | 7623 | 9053125 | ||
BC150625.1 | - | 38 | 7623 | 9056995 | ||
BC171790.1 | - | 38 | 7623 | 9054444 | ||
BX641155.1 | - | 18 | 5620 | |||
X52022.1 | - | 41 | 9470 |