Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other MTUS1 SIRNA Assays ›
Gene Symbol:
MTUS1
Gene Name:
microtubule associated tumor suppressor 1
Gene Aliases:
ATBP, ATIP, ICIS, MP44, MTSG1
Chromosome Location:
Chr.8: 17643794 - 17801579 on Build GRCh38
UniGene ID:
Hs.7946
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s33189
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: MTUS1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001001924.2
NP_001001924.1
7
3192
NM_001001925.2
NP_001001925.1
6
3030
NM_001001931.2
NP_001001931.1
4
779
NM_001166393.1
NP_001159865.1
4
424
NM_020749.4
NP_065800.1
2
576
XM_005273577.3
XP_005273634.1
7
3104
XM_005273578.3
XP_005273635.1
7
3192
XM_005273579.3
XP_005273636.1
7
3104
XM_005273580.3
XP_005273637.1
6
2942
XM_005273581.3
XP_005273638.1
6
3030
XM_005273582.3
XP_005273639.1
6
2942
XM_005273583.4
XP_005273640.1
7
3104
XM_011544595.2
XP_011542897.1
7
3104
XM_017013698.1
XP_016869187.1
8
3110
XM_017013699.1
XP_016869188.1
8
3332
XM_017013700.1
XP_016869189.1
7
2991
XM_017013701.1
XP_016869190.1
8
3108
XM_017013702.1
XP_016869191.1
6
2763
GenBank mRNA
AB033114.1
-
6
2644
AF121259.1
-
2
216
AF293357.1
-
2
489
AK024357.1
-
1
102
AK125188.1
-
4
779
AK289750.1
-
2
575
AK294860.1
-
4
365
AK304105.1
-
5
661
AK314692.1
-
2
510
AL096842.1
-
7
1984
BC007328.2
-
6
678
BC117564.1
-
2
254
BC136320.1
-
7
2943
9020307
BC142971.1
-
2
509
DA113649.1
-
4
425
Back To Top
More Information
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5d97d974fc-r4vqj:80/10.239.163.135:80.
git-commit: fe8b04579249ff98075bad6061a6e2a4739e01ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.13.0-2024.08.32-1.0