Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CC2D2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080522.2 | NP_001073991.2 | 8 | 757 | ||
XM_005248177.1 | XP_005248234.1 | 7 | 712 | |||
XM_011513872.2 | XP_011512174.1 | 7 | 712 | |||
XM_017008482.1 | XP_016863971.1 | 5 | 506 | |||
GenBank mRNA | AB037766.1 | - | 5 | 495 | ||
AK096047.1 | - | 7 | 699 | |||
AK309178.1 | - | 7 | 687 | |||
BC103710.1 | - | 5 | 485 | |||
BX647334.1 | - | 7 | 692 | |||
DB043556.1 | - | 5 | 506 | |||
EU450799.1 | - | 8 | 584 |