Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NCEH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001146276.1 | NP_001139748.1 | 5 | 1060 | ||
NM_001146277.1 | NP_001139749.1 | 5 | 1026 | |||
NM_001146278.1 | NP_001139750.1 | 4 | 807 | |||
NM_020792.4 | NP_065843.3 | 5 | 1036 | |||
GenBank mRNA | AB037784.1 | - | 5 | 869 | ||
AK294811.1 | - | 5 | 879 | |||
AK295641.1 | - | 4 | 650 | |||
AK299422.1 | - | 5 | 908 | |||
AK304253.1 | - | 5 | 937 | |||
AK316431.1 | - | 5 | 931 | |||
BC028734.1 | - | 5 | 1041 | 4815834 | ||
BC047588.1 | - | 5 | 885 |