Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: USP35 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020798.2 | NP_065849.1 | 5 | 1341 | ||
XM_011545187.2 | XP_011543489.1 | 5 | 484 | |||
XM_011545188.2 | XP_011543490.1 | 3 | 267 | |||
XM_017018052.1 | XP_016873541.1 | 5 | 1338 | |||
XM_017018053.1 | XP_016873542.1 | 4 | 333 | |||
GenBank mRNA | AB037793.1 | - | 2 | 804 | ||
AJ586137.1 | - | 4 | 982 | |||
AK307875.1 | - | 4 | 567 | |||
BC131489.1 | - | 3 | 304 | 40038029 |