Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PREX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020820.3 | NP_065871.2 | 35 | 4532 | ||
XM_011528934.2 | XP_011527236.1 | 27 | 3434 | |||
GenBank mRNA | AB037836.2 | - | 35 | 4395 | ||
AJ320261.1 | - | 35 | 4532 | |||
AL136579.1 | - | 18 | 2489 | |||
AL832913.1 | - | 28 | 3501 | |||
BC053616.1 | - | 31 | 3982 |