Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM181 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
RefSeq | NM_020823.1 | NP_065874.1 | 5 | 714 | ||
XM_005267074.3 | XP_005267131.1 | 5 | 431 | |||
XM_011535999.1 | XP_011534301.1 | 5 | 718 | |||
XM_011536000.1 | XP_011534302.1 | 5 | 718 | |||
XM_011536001.2 | XP_011534303.1 | 5 | 898 | |||
XM_011536002.2 | XP_011534304.1 | 5 | 896 | |||
XM_011536003.2 | XP_011534305.1 | 5 | 718 | |||
GenBank mRNA | AB037844.1 | - | 5 | 716 | ||
AY927471.1 | - | LIT3489 AY927471 |
5 | 831 | ||
AY927472.1 | - | 6 | 1367 | |||
BC033450.1 | - | 4 | 437 |