Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WDFY1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020830.4 | NP_065881.1 | 5 | 458 | ||
GenBank mRNA | AB037856.1 | - | 5 | 371 | ||
AF411977.1 | - | 5 | 355 | |||
AJ310568.1 | - | 5 | 445 | |||
AK022888.1 | - | 5 | 384 | |||
AK023415.1 | - | 4 | 252 | |||
BC040525.1 | - | 5 | 370 | 5285567 | ||
BC065934.1 | - | 5 | 402 | 6164567 | ||
DA775670.1 | - | 5 | 458 |