Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CALCOCO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001143682.1 | NP_001137154.1 | 9 | 1223 | ||
NM_020898.2 | NP_065949.1 | 10 | 1478 | |||
NR_026554.1 | 9 | 1375 | ||||
XM_011538601.1 | XP_011536903.1 | 10 | 1412 | |||
XM_011538602.1 | XP_011536904.1 | 11 | 1545 | |||
XM_011538603.1 | XP_011536905.1 | 11 | 1563 | |||
XM_011538604.1 | XP_011536906.1 | 11 | 1519 | |||
XM_017019707.1 | XP_016875196.1 | 11 | 1545 | |||
XM_017019708.1 | XP_016875197.1 | 9 | 1540 | |||
XM_017019709.1 | XP_016875198.1 | 9 | 1540 | |||
GenBank mRNA | AB040969.1 | - | 10 | 1375 | ||
AF370415.1 | - | 10 | 1383 | |||
AK027881.1 | - | 10 | 1412 | |||
AK092788.1 | - | 9 | 1222 | |||
AK122773.1 | - | 9 | 1139 | |||
AK294424.1 | - | 10 | 1386 | |||
AK315578.1 | - | 10 | 1413 | |||
AL136895.1 | - | 10 | 1390 | |||
AY211909.1 | - | 6 | 897 | |||
AY358397.1 | - | 10 | 1387 | |||
AY563137.1 | - | 10 | 1385 | |||
BC003177.1 | - | 10 | 1385 | 2957946 | ||
CR533567.1 | - | 9 | 1334 |