Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLEKHA4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001161354.1 | NP_001154826.1 | 5 | 867 | ||
NM_020904.2 | NP_065955.2 | 5 | 867 | |||
XM_005259107.2 | XP_005259164.1 | 5 | 862 | |||
XM_006723301.2 | XP_006723364.1 | 5 | 862 | |||
XM_011527157.1 | XP_011525459.1 | 5 | 862 | |||
XM_011527158.2 | XP_011525460.1 | 4 | 701 | |||
XM_011527159.1 | XP_011525461.1 | 5 | 875 | |||
XM_011527160.1 | XP_011525462.1 | 5 | 862 | |||
XM_017027037.1 | XP_016882526.1 | 2 | 139 | |||
GenBank mRNA | AB208908.1 | - | 5 | 829 | ||
AY007233.1 | - | 5 | 841 | |||
BC024157.2 | - | 5 | 847 | 3953478 | ||
BC064601.1 | - | 5 | 694 | 5785991 |