Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZDBF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001285549.1 | NP_001272478.1 | 7 | 1503 | ||
NM_020923.2 | NP_065974.1 | 5 | 1610 | |||
XM_005246711.3 | XP_005246768.1 | 6 | 1610 | |||
XM_005246712.4 | XP_005246769.1 | 6 | 1433 | |||
XM_005246713.3 | XP_005246770.1 | 6 | 1427 | |||
XM_006712656.3 | XP_006712719.1 | 6 | 1466 | |||
XM_011511532.2 | XP_011509834.1 | 6 | 1643 | |||
XM_011511533.2 | XP_011509835.1 | 6 | 1460 | |||
XM_011511534.2 | XP_011509836.1 | 5 | 1383 | |||
XM_011511535.2 | XP_011509837.1 | 5 | 1389 | |||
XM_017004573.1 | XP_016860062.1 | 7 | 1716 | |||
XM_017004574.1 | XP_016860063.1 | 7 | 1642 | |||
XM_017004575.1 | XP_016860064.1 | 6 | 1611 | |||
GenBank mRNA | AB775780.1 | - | 7 | 1504 | ||
AB775781.1 | - | 5 | 1610 | |||
AK127271.1 | - | 5 | 1455 |