Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZSWIM6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020928.1 | NP_065979.1 | 10, 11 | 2382 | ||
XM_017009677.1 | XP_016865166.1 | 8, 9 | 1333 | |||
GenBank mRNA | AB046797.1 | - | 6, 7 | 968 |