Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC4A5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021196.3 | NP_067019.3 | 18 | 2571 | ||
NM_133478.2 | NP_597812.1 | 23 | 2967 | |||
GenBank mRNA | AB209752.1 | - | 19 | 3374 | ||
AF243499.2 | - | 18 | 2569 | |||
AF293337.2 | - | 22 | 2916 | |||
AF293338.2 | - | 18 | 2569 | |||
AF452248.1 | - | 20 | 2750 | |||
AF453528.1 | - | 19 | 2644 | |||
BC109221.2 | - | 17 | 2312 | 40008543 |