Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MS4A7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021201.4 | NP_067024.1 | 5 | 678 | ||
NM_206938.1 | NP_996821.1 | 4 | 543 | |||
NM_206939.1 | NP_996822.1 | 5 | 630 | |||
NM_206940.1 | NP_996823.1 | 4 | 495 | |||
GenBank mRNA | AB026043.1 | - | 5 | 618 | ||
AF201951.1 | - | 5 | 589 | |||
AF237916.1 | - | 5 | 602 | |||
AF309653.1 | - | 5 | 631 | |||
AF350501.1 | - | 5 | 589 | |||
AK075106.1 | - | 5 | 682 | |||
AK297027.1 | - | 4 | 479 | |||
AK309884.1 | - | 4 | 496 | |||
BC020673.1 | - | 5 | 602 | 4720044 | ||
CR457187.1 | - | 4 | 485 |