Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABHD17C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021214.1 | NP_067037.1 | 2 | 732 | ||
GenBank mRNA | AK092758.1 | - | 2 | 810 | ||
AK298606.1 | - | 2 | 318 | |||
AL390079.1 | - | 2 | 353 | |||
BC059401.1 | - | 2 | 702 | 30346383 | ||
BC115003.1 | - | 2 | 732 | 7503983 |