Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRUNE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001303229.1 | NP_001290158.1 | 2 | 365 | ||
NM_001303242.1 | NP_001290171.1 | 2 | 365 | |||
NM_001303243.1 | NP_001290172.1 | 2 | 365 | |||
NM_021222.2 | NP_067045.1 | 2 | 365 | |||
XM_005245393.4 | XP_005245450.1 | 2 | 343 | |||
XM_017001955.1 | XP_016857444.1 | 2 | 343 | |||
XM_017001957.1 | XP_016857446.1 | 2 | 344 | |||
GenBank mRNA | AF051907.1 | - | 2 | 279 | ||
AK315120.1 | - | 2 | 296 | |||
BC014886.1 | - | 2 | 273 | 3907577 | ||
BC025304.1 | - | 2 | 259 | 5458046 | ||
BM554427.1 | - | 2 | 279 | |||
DA719664.1 | - | 2 | 365 | |||
U67085.1 | - | 1 | 71 |