Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF462 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021224.4 | NP_067047.4 | 3 | 2969 | ||
XM_006717209.3 | XP_006717272.1 | 3 | 2965 | |||
XM_006717211.3 | XP_006717274.1 | 3 | 4011 | |||
XM_006717212.3 | XP_006717275.1 | 3 | 2770 | |||
XM_006717215.3 | XP_006717278.1 | 3 | 4760 | |||
XM_006717216.3 | XP_006717279.1 | 3 | 4755 | |||
XM_006717218.3 | XP_006717281.1 | 3 | 4687 | |||
XM_011518892.2 | XP_011517194.1 | 3 | 4760 | |||
XM_017014996.1 | XP_016870485.1 | 3 | 3112 | |||
XM_017014997.1 | XP_016870486.1 | 3 | 2927 | |||
XM_017014998.1 | XP_016870487.1 | 3 | 4685 | |||
XM_017014999.1 | XP_016870488.1 | 3 | 2965 | |||
GenBank mRNA | AK127803.1 | - | 3 | 2965 | ||
BX648965.1 | - | 3 | 2969 | |||
CR749850.1 | - | 1 | 2371 |