Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COX5B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001862.2 | NP_001853.2 | 2, 3 | 204 | ||
GenBank mRNA | BC006229.2 | - | 2, 3 | 204 | 3952882 | |
BT006742.1 | - | 2, 3 | 175 | |||
CR541727.1 | - | 2, 3 | 175 | |||
CR541753.1 | - | 2, 3 | 175 | |||
M19961.1 | - | 2, 3 | 196 |