Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COX6C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004374.3 | NP_004365.1 | 3 | 577 | ||
XM_017013020.1 | XP_016868509.1 | 3 | 383 | |||
GenBank mRNA | AK311791.1 | - | 3 | 270 | ||
BC000187.2 | - | 2 | 202 | 3350637 | ||
BT007007.1 | - | 2 | 170 | |||
S82616.1 | - | 1 | 52 | |||
X13238.1 | - | 3 | 237 |