Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SCPEP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021626.2 | NP_067639.1 | 5 | 585 | ||
XM_005257557.2 | XP_005257614.1 | 5 | 567 | |||
GenBank mRNA | AF113214.1 | - | 5 | 510 | ||
AF282618.1 | - | 5 | 564 | |||
AK027373.1 | - | 5 | 558 | |||
AK092928.1 | - | 2 | 822 | |||
AK298161.1 | - | 5 | 504 | |||
AY358559.1 | - | 5 | 565 | |||
BC072405.1 | - | 5 | 535 | 6137668 |