Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COX7B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001866.2 | NP_001857.1 | 2 | 190 | ||
GenBank mRNA | AK311879.1 | - | 2 | 172 | ||
BC018386.1 | - | 2 | 173 | 3861730 | ||
BT009767.1 | - | 2 | 100 | |||
CR450332.1 | - | 2 | 100 | |||
CR542124.1 | - | 2 | 100 | |||
Z14244.1 | - | 2 | 202 |