Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AFAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134647.1 | NP_001128119.1 | 6 | 879 | ||
NM_198595.2 | NP_940997.1 | 6 | 879 | |||
XM_006713909.3 | XP_006713972.1 | 6 | 762 | |||
XM_011513544.2 | XP_011511846.1 | 6 | 932 | |||
XM_017008535.1 | XP_016864024.1 | 5 | 608 | |||
XM_017008536.1 | XP_016864025.1 | 5 | 608 | |||
GenBank mRNA | AB209676.1 | - | 3 | 332 | ||
AF188700.1 | - | 5 | 606 | |||
AK290807.1 | - | 6 | 706 | |||
AK297631.1 | - | 6 | 672 | |||
BC032777.1 | - | 5 | 630 | 4914388 | ||
CN308844.1 | - | 3 | 358 |