Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC90B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286116.1 | NP_001273045.1 | 5 | 663 | ||
NM_001286117.1 | NP_001273046.1 | 5 | 1210 | |||
NM_001286118.1 | NP_001273047.1 | 4 | 786 | |||
NM_001286119.1 | NP_001273048.1 | 5 | 871 | |||
NM_001286120.1 | NP_001273049.1 | 3 | 666 | |||
NM_021825.4 | NP_068597.2 | 5 | 890 | |||
XM_005274154.2 | XP_005274211.1 | 4 | 1045 | |||
XM_005274156.3 | XP_005274213.1 | 5 | 877 | |||
XM_005274159.3 | XP_005274216.1 | 6 | 556 | |||
XM_017018103.1 | XP_016873592.1 | 4 | 488 | |||
XM_017018104.1 | XP_016873593.1 | 6 | 981 | |||
GenBank mRNA | AF182412.1 | - | 5 | 460 | ||
AF182424.1 | - | 5 | 460 | |||
AF271782.1 | - | 5 | 817 | |||
AK055972.1 | - | 5 | 1207 | |||
AK225726.1 | - | 5 | 471 | |||
AK304769.1 | - | 5 | 623 | |||
AK315787.1 | - | 5 | 851 | |||
AL136791.1 | - | 5 | 610 | |||
AV756714.1 | - | 4 | 426 | |||
BC014573.1 | - | 5 | 500 | 3680689 | ||
BC020783.1 | - | 5 | 461 | 4773466 | ||
BC067764.1 | - | 5 | 781 | |||
BC107755.1 | - | 5 | 631 | 6737596 | ||
BG700162.1 | - | 3 | 243 |