Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GUF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021927.2 | NP_068746.2 | 15 | 1991 | ||
XM_005248122.3 | XP_005248179.1 | 14 | 1845 | |||
GenBank mRNA | AK023282.1 | - | 15 | 1941 | ||
AK025248.1 | - | 7 | 771 | |||
AK225314.1 | - | 15 | 1946 | |||
BC036768.1 | - | 15 | 1952 | 5744283 |