Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPCS3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021928.3 | NP_068747.1 | 4 | 503 | ||
GenBank mRNA | AK026302.1 | - | 4 | 478 | ||
AK092634.1 | - | 3 | 1486 | |||
AL136660.1 | - | 4 | 473 | |||
AY359044.1 | - | 4 | 461 | |||
BC047290.1 | - | 4 | 446 | 5224927 | ||
BC053883.1 | - | 3 | 2397 | |||
DB285025.1 | - | 4 | 505 |