Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RINT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021930.4 | NP_068749.3 | 10 | 1634 | ||
XM_005250524.3 | XP_005250581.1 | 6 | 816 | |||
XM_011516458.2 | XP_011514760.1 | 6 | 789 | |||
XM_017012496.1 | XP_016867985.1 | 10 | 1413 | |||
XM_017012497.1 | XP_016867986.1 | 10 | 1505 | |||
XM_017012498.1 | XP_016867987.1 | 7 | 1028 | |||
GenBank mRNA | AF317622.1 | - | 10 | 1513 | ||
AK021847.1 | - | 2 | 306 | |||
AK055508.1 | - | 10 | 1549 | |||
AK057757.1 | - | 7 | 1035 | |||
AK308126.1 | - | 5 | 427 | |||
AL832825.1 | - | 4 | 462 | |||
BC068483.1 | - | 10 | 1635 | 5270834 |