Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZFAND3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021943.2 | NP_068762.1 | 3 | 684 | ||
XM_005249269.1 | XP_005249326.1 | 3 | 719 | |||
XM_011514790.1 | XP_011513092.1 | 3 | 719 | |||
XM_017011171.1 | XP_016866660.1 | 2 | 657 | |||
GenBank mRNA | AK023284.1 | - | 3 | 358 | ||
BC031481.2 | - | 3 | 300 | 4448968 | ||
BP305887.1 | - | 3 | 413 |