Gene Symbol: PKNOX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022062.2 | NP_071345.2 | 9 | 1046 | ||
XM_005271642.2 | XP_005271699.1 | 8 | 935 | |||
XM_005271643.2 | XP_005271700.1 | 6 | 752 | |||
XM_006718894.2 | XP_006718957.1 | 8 | 822 | |||
XM_011542944.2 | XP_011541246.1 | 9 | 1131 | |||
XM_011542945.2 | XP_011541247.1 | 10 | 1091 | |||
XM_011542947.2 | XP_011541249.1 | 7 | 700 | |||
XM_017018110.1 | XP_016873599.1 | 9 | 3881 | |||
XM_017018111.1 | XP_016873600.1 | 7 | 741 | |||
GenBank mRNA | AB065001.1 | - | 6 | 724 | ||
AJ417081.1 | - | 6 | 696 | |||
AK023136.1 | - | 6 | 724 | |||
AK295857.1 | - | 8 | 913 | |||
AK298999.1 | - | 8 | 815 | |||
AK316001.1 | - | 8 | 846 | |||
AK316268.1 | - | 7 | 834 | |||
AL512682.1 | - | 3 | 979 | |||
BC045626.1 | - | 9 | 1003 | 5261106 | ||
BX648615.1 | - | 7 | 823 |