Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RNF123 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022064.4 | NP_071347.2 | 35 | 3578 | ||
NR_135218.1 | 35 | 3818 | ||||
XM_006713285.1 | XP_006713348.1 | 35 | 3736 | |||
XM_011533995.1 | XP_011532297.1 | 35 | 3513 | |||
XM_017007018.1 | XP_016862507.1 | 35 | 3540 | |||
GenBank mRNA | AF370367.1 | - | 26 | 2732 | ||
AK022627.1 | - | 13 | 1404 | |||
AK026968.1 | - | 17 | 1832 | |||
AK090632.1 | - | 25 | 2620 | |||
AK096353.1 | - | 35 | 3492 | |||
AK126883.1 | - | 19 | 3285 | |||
AL136729.1 | - | 8 | 962 | |||
AL832034.1 | - | 35 | 3540 | |||
AL832138.1 | - | 34 | 3829 | |||
AL832344.1 | - | 35 | 3516 | |||
AY744152.1 | - | 34 | 3461 | |||
BC041145.1 | - | 20 | 2111 | |||
BC057392.1 | - | 22 | 2268 | |||
BC088801.1 | - | 17 | 1839 | |||
BC130632.1 | - | 35 | 3519 | 40146663 | ||
CR627051.1 | - | 31 | 5150 |